missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ GHB sodium salt Solution, 1.0 mg/mL in Methanol (as salt), Sold by MilliporeSigma™ Supelco™
Description
GHB, or γ-hydroxybutyrate, is a central nervous system depressant sold under the brand name Xyrem™ and used medically to treat narcolepsy, excessive daytime sleepiness and in rare circumstances, alcoholism. GHB is also used recreationally as an intoxicant and date rape drug. Its many street names include 'GBH', 'Liquid X', 'G', 'Juice', and 'Mils'.
Specifications
Specifications
| CAS | 67-56-1, 502-85-2 |
| Linear Formula | C4H7NaO3 |
| MDL Number | MFCD00004402 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol (as salt) |
| Synonym | 4-Hydroxybutyric acid sodium salt solution |
| Formula Weight | 126.09 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation