missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Furanyl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001370
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001370 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001370 Supplier Merck Emd Millipore Supplier No. F0460.5ML

Certified reference material

An opioid analgesic and fentanyl analog with about one fifth the potency of fentanyl. Furanyl fentanyl, or Fu-F, is sold on the illicit market as a designer drug or in combination with heroin. This Certified Spiking Solution™ is suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C24H26N2O2 · HCl
MDL Number MFCD30718274
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 410.94

CERILLIANT is a registered trademark of Cerilliant Corporation