missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Furanyl fentanyl-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
Furanyl fentanyl, or Fu-F, is sold on the illicit drug market as a designer drug or in combination with heroin. This stable-labeled internal standard is suitable for quantitation of furanyl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, urine drug testing, forensic analysis, or isotope dilution methods. This Certified Spiking Solution™ is supplied in a convenient US DEA-exempt solution format.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Concentration | 100 μg/mL in methanol (as free base) |
| Linear Formula | C24H21D5N2O2 · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 0.5 mL |
| Formula Weight | 415.97 |
CERILLIANT is a registered trademark of Cerilliant Corporation