Learn More
CERILLIANT™ Fluconazole-13C3 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore F0351ML
Description
A stable-labeled internal standard suitable for quantification of fluconazole levels in blood, serum, or plasma by HPLC or LC-MS/MS. Fluconazole, sold under trade names such as Trican, Monicure, Diflucan™, and Afungil, is an antifungal drug used in the treatment and prevention of superficial and systemic fungal infections.Specifications
| 67-56-1 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 309.25 | |
| 1 mL |
| C1013C3H12F2N6O | |
| 1.0 mg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation