missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Fentanyl Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001346
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001346 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001346 Supplier Merck Emd Millipore Supplier No. F0021ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material

A potent synthetic narcotic analgesic and local anesthetic with a rapid onset and short duration of action. Sold under various trade names including Sublimaze, Duragesic™, Actiq™, and Fentora™, fentanyl is more potent than morphine in analgesic activity. This certified solution standard is suitable for a variety of LC/MS or GC/MS testing applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C22H28N2O
MDL Number MFCD00661055
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 336.47

CERILLIANT is a registered trademark of Cerilliant Corporation