missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Fentanyl-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001345
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001345 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001345 Supplier Merck Emd Millipore Supplier No. F0011ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material

An internal standard suitable for fentanyl testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring, or forensic analysis. Fentanyl is a potent synthetic narcotic analgesic and local anesthetic sold under various trade names including Sublimaze, Duregesic, Actiq™, and Fentora™.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C22D5H23N2O
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 341.5

CERILLIANT is a registered trademark of Cerilliant Corporation