missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Everolimus Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Everolimus, sold under trade names including Zortress™, Certican, and Afinitor™, is an immunosuppressant drug used to prevent rejection of organ transplants and to treat renal cell cancer and other tumors. This Certified Spiking Solution™ is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of everolimus in patient whole blood samples by LC-MS/MS.
Specifications
Specifications
| CAS | 75-05-8, 159351-69-6 |
| Linear Formula | C53H83NO14 |
| MDL Number | MFCD00929329 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 958.22 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation