missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Everolimus-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
A stable-labeled internal standard for the immunosuppressant Everolimus, marketed as Zortress™ and Certican for use in organ transplantation and Afinitor™ for treatment of kidney and pancreatic cancers. Clinical laboratories quantitate everolimus levels in patient whole blood samples using LC-MS/MS. While many everolimus LC-MS/MS methods use 32-desmethoxyrapamycin or ascomycin as an internal standard, higher levels of variability / lower levels of reproducibility may occur in the analysis along with the potential for interferences. Use of a stable-labeled analog of everolimus is the best choice to minimize variability or mitigate any potential interference in LC-MS/MS analysis.
Specifications
Specifications
| CAS | 75-05-8, 1338452-54-2 |
| Linear Formula | C53H79NO14D4 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 962.25 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation