missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Eutylone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
Eutylone is a synthetic cathinone found in numerous bath salt blends under the street name bk-MBDB. This designer drug and stimulant is abused in the US and Europe. This certified Snap-N-Spike™ solution is suitable for quantification of eutylone using LC/MS or GC/MS methods for applications in clinical toxicology, forensic analysis, and urine drug testing applications.
Specifications
Specifications
| CAS | 67-56-1, 17764-18-0 |
| Linear Formula | C13H17NO3 · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Formula Weight | 271.74 |
| Packaging | Ampule |
| Quantity | 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation