missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ethylone Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001298
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001298 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001298 Supplier Merck Emd Millipore Supplier No. E0711ML

Certified reference material

Also known as bk-MDEA or 3,4-methylenedioxy-N-ethylcathinone, ethylone is a designer drug of the amphetamine and cathinone classes with cases of abuse reported in Europe. This certified solution standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology testing and forensic analysis to urine drug testing.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C12H15NO3 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym Ethylone hydrochloride solution
Quantity 1 mL
Formula Weight 257.71

CERILLIANT is a registered trademark of Cerilliant Corporation