missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ethylone-d5 Hydrochloride, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001299
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001299 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001299 Supplier Merck Emd Millipore Supplier No. E0721ML
Only null left

Certified reference material

A new internal standard for the quantitation of ethylone in biological samples using LC/MS and GC/MS. Also known as bk-MDEA or 3,4-methylenedioxy-N-ethylcathinone, ethylone is a designer drug of the amphetamine and cathinone classes with cases of abuse reported in Europe. This certified solution standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology testing and forensic analysis to urine drug testing and isotope dilution methods.

Specifications

CAS 67-56-1
Linear Formula C12D5H10NO3 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym Ethylone-D5 hydrochloride solution
Formula Weight 262.74
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation