Learn More
CERILLIANT™ Ethylmorphine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore E0521ML
Description
Ethylmorphine is an opioid analgesic metabolized by the liver into morphine. The drug carries a risk of dependence and abuse. This certified solution standard is suited for LC/MS or GC/MS methods in forensic analysis, clinical toxicology, and urine drug testing.Specifications
| 67-56-1,76-58-4 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 313.39 | |
| 1 mL |
| C19H23NO3 | |
| 1.0 mg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation