missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ethyl sulfate sodium salt, 1.0 mg/mL in Methanol (as ethyl sulfate), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001294
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001294 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001294 Supplier Merck Emd Millipore Supplier No. E0641ML

Certified reference material

A Certified Spiking Solution™ suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine alcohol testing. Ethyl sulfate is a primary urinary metabolite of consumed alcohol and is indicative of recent alcohol consumption.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as ethyl sulfate)
Linear Formula C2H5O4SNa
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 148.11

CERILLIANT is a registered trademark of Cerilliant Corporation