missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ethyl-d5 sulfate sodium salt, 1.0 mg/mL in Methanol (as ethyl sulfate), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001295
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001295 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001295 Supplier Merck Emd Millipore Supplier No. E0661ML

Certified reference material

Ethyl sulfate is a metabolite of ethanol in humans and a biomarker of acute alcohol consumption screened in blood alcohol testing. This new internal standard is suitable for a variety of uses in LC/MS or GC/MS applications from blood alcohol/ethanol testing and forensic or clinical toxicology analysis to urine drug testing.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as ethyl sulfate)
Linear Formula C2D5NaO4S
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym Ethyl-d5 sulfate sodium salt
Quantity 1 mL
Formula Weight 153.14

CERILLIANT is a registered trademark of Cerilliant Corporation