missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ethyl-beta-D-glucuronide-(ethyl-d5), 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001286
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001286 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001286 Supplier Merck Emd Millipore Supplier No. E0481ML

Certified reference material

Ethyl β-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alcohol consumption. This internal standard is suitable for quantitation of alcohol levels in the body by LC/MS or GC/MS for applications such as forensic testing, clinical toxicology, or alcohol monitoring.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C8D5H9O7
MDL Number MFCD09037575
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 227.22

CERILLIANT is a registered trademark of Cerilliant Corporation