missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ EDDP-D3 perchlorate Solution, 100 μg/mL in Methanol (as pyrrolinium), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001281
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001281 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001281 Supplier Merck Emd Millipore Supplier No. E0211ML

Certified reference material

EDDP is the major urinary metabolite of methadone, a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. This stable-labeled internal standard is suitable for EDDP and methadone testing methods by GC/MS or LC/MS for forensic analysis, urine drug testing or clinical toxicology.

Specifications

Packaging Ampule
CAS 67-56-1, 136765-23-6
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as pyrrolinium)
Linear Formula C20D3H21N · ClO4
MDL Number MFCD00673273
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym EDDP-D3 perchlorate solution, Methadone metabolite
Quantity 1 mL
Formula Weight 380.88

CERILLIANT is a registered trademark of Cerilliant Corporation