missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Diclazepam-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001239
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001239 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001239 Supplier Merck Emd Millipore Supplier No. D1601ML

Certified reference material

Diclazepam, also known as, chlorodiazepam and 2′-chloro-diazepam, is a benzodiazepine and functional analog of diazepam that has emerged on the designer drug market. This stable-labeled internal standard is suitable for quantification of diclazepam by LC/MS or GC/MS for clinical toxicology, forensic analysis, prescription monitoring, or urine drug testing applications.

Specifications

CAS 75-05-8, 84344-18-3
Linear Formula C16H8Cl2D4N2O
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 323.21
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation