missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Dibenz[a,h]anthracene, , Sold by MilliporeSigma™ Supelco™

Catalog No. 01002024
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002024 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002024 Supplier Merck Emd Millipore Supplier No. SCD0031G
Only null left

Certified reference material

Dibenz[a,h]anthracene belongs to the class of polynuclear aromatic hydrocarbons (PAHs). PAHs are a group of well-known environmental pollutants with both mutagenic and carcinogenic properties. They are released into the environment by the pyrolysis of organic materials.

Specifications

CAS 53-70-3
Boiling Point 524°C (lit.)
Melting Point 262 to 265°C (lit.)
Linear Formula C22H14
MDL Number MFCD00003708
UN Number UN 3077 9 / PGIII
Synonym 1,2:5,6-Dibenzanthracene
Formula Weight 278.35
Packaging Vial
Quantity 1 g

CERILLIANT is a registered trademark of Cerilliant Corporation