missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ (+/-)-Delta9-THC Solution, 100 μg/mL in heptane, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002057
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002057 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002057 Supplier Merck Emd Millipore Supplier No. T0471ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material

(±)-δ9-THC is the main active constituent and the primary psychoactive cannabinoid of cannabis. Besides persistent illicit use this drug is suitable as treatment for anorexia and nausea and is sold under the trade name Marinol™. This Certified Spiking Solution™ is suitable as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Specifications

CAS 142-82-5, 6465-30-1
Linear Formula C21H30O2
UN Number UN 1206 - class 3 - PG 2 - Heptanes, solution
Concentration 100 μg/mL in heptane
Formula Weight 314.46
Packaging Ampule
Quantity 1 mL
Flash Point -4°C (24.8°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation