missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ D6-25-Hydroxyvitamin D3 (26,26,26,27,27,27-D6) Solution, 50 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001437
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001437 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001437 Supplier Merck Emd Millipore Supplier No. H0741ML

Certified reference material

A Certified Spiking Solution of D6-25-Hydroxyvitamin D3 (26,26,26,27,27,27-D6) in ethanol at a concentration of 50 μg/mL. This new internal standard is suitable for critical quantitative applications in research and clinical environments including for use in preparation of calibrators for LC/UV and LC-MS/MS assays and isotope dilution methods. The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike™ format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Specifications

CAS 64-17-5, 78782-98-6
Linear Formula C27H38D6O2
MDL Number MFCD15071183
UN Number UN 1170 - class 3 - PG 2 - Ethanol, solution
Concentration 50 μg/mL in ethanol
Formula Weight 406.67
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation