Learn More
CERILLIANT™ Cyclosporin A Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore C0931ML
Catalog No. 01001069
Only null left
Explore available promotions
Description
Description
A Certified Spiking Solution™ of an immunosuppressant and narrow therapeutic index drug used to prevent rejection in organ transplantation. This certified calibration material is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the drug's narrow therapeutic range. The Snap-N-Spike™ format allows laboratories to eliminate weighing operations of hazardous substances such as cyclosporin A, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.
Specifications
Specifications
75-05-8,59865-13-3
UN 1648 3 / PGII
1202.61
1 mL
C62H111N11O12
1.0 mg/mL in acetonitrile
Ampule
2°C (35.6°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation