missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Cyclopropyl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001129
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001129 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001129 Supplier Merck Emd Millipore Supplier No. C1770.5ML

Certified reference material

Cyclopropyl fentanyl is an opioid analgesic analog of fentanyl and has been sold as a designer drug on the illicit drug market. Cyclopropyl fentanyl is a Schedule I controlled drug in the USA since January 2018 and was banned in Finland and Sweden in 2017. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. This Certified Spiking Solution™ is supplied in a convenient US DEA-exempt solution format.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C23H28N2O · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 384.94

CERILLIANT is a registered trademark of Cerilliant Corporation