missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Cortisol-D4 (9,11,12, 12-D4) Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C1131ML
Description
Cortisol, also known as hydrocortisone, is a glucocorticoid produced by the adrenal gland. This stable labeled internal standard is suitable for quantitation of cortisol levels in whole blood by LC-MS/MS for newborn screening of congenital adrenal hyperplasia (CAH) and in serum or plasma by LC-MS/MS for determination of several diseases including Cushing's syndrome, Addison's disease, and Sheehan's syndrome.Specifications
| 67-56-1,73565-87-4 | |
| MFCD10567120 | |
| 100 μg/mL in methanol | |
| 366.48 | |
| 1 mL |
| C21H26D4O5 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| Cortisol-D4 (9,11,12,12-D4) solution, Hydrocortisone-9,11,12,12-D4 solution | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation