missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Coenzyme Q10 Solution, 1 mg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002174
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002174 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002174 Supplier Merck Emd Millipore Supplier No. V0601ML

Certified reference material

Coenzyme Q10, also known as ubiquinone, ubidecarenone, coenzyme Q, CoQ10, CoQ or Q10, is a fat soluble cofactor, which resembles a vitamin and is important to human health as an antioxidant.

Specifications

CAS 64-17-5, 303-98-0
Linear Formula C59H90O4
MDL Number MFCD00042919
UN Number UN 1170 - class 3 - PG 2 - Ethanol, solution
Concentration 1 mg/mL in ethanol
Formula Weight 863.34
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation