missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Codeine-6-ß-D-glucuronide-D3 Solution, 100 μg/mL in Methanol:Water (2:8), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001097
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001097 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001097 Supplier Merck Emd Millipore Supplier No. C1381ML
Only null left

Certified reference material

An internal standard suitable for LC/MS or GC/MS applications with codeine-6β-D-glucuronide such as forensic analysis, urine drug testing or clinical toxicology. Codeine-6β-D-glucuronide is a major urinary metabolite of codeine, an opiate commonly used as an antitussive and analgesic for management of mild pain. This Certified Spiking Solution™ provides added value in allowing for quantitation of codeine-6β-D-glucuronide levels by LC-MS/MS without the need for hydrolysis.

Specifications

CAS 67-56-1, 219533-59-2
Linear Formula C24H26D3NO9
Concentration 100 μg/mL in methanol:water (2:8)
Formula Weight 478.51
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation