Learn More
CERILLIANT™ Clorazepate dipotassium Solution, 1.0 mg/mL in Methanol (as clorazepate), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C1561ML
Description
A certified analytical reference solution suitable for use in benzodiazepine testing methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing or forensic analysis. Clorazepate dipotassium is a long-acting benzodiazepine with anxiolytic, anticonvulsant, sedative and hypnotic properties. Clorazepate dipotassium, marketed under the brand name Tranxene™, is a prodrug for nordiazepam, which is produced as an active metabolite.Specifications
| 67-56-1,57109-90-7 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 408.92 | |
| 1 mL |
| C16H11ClK2N2O4 | |
| 1.0 mg/mL in methanol (as clorazepate) | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation