missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Clonazolam Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
A certified solution standard for use in LC/MS or GC/MS benzodiazepine testing methods in clinical toxicology, urine drug testing, or forensic analysis. Clonazolam, also known as clonitrazolam, is a highly potent benzodiazepine analog that can cause strong sedation and amnesia at minimal oral doses. Clonazolam is also sold on the designer drug market with cases of abuse noted in Europe and the US.
Specifications
Specifications
| CAS | 75-05-8, 33887-02-4 |
| Linear Formula | C17H12ClN5O2 |
| MDL Number | MFCD30718196 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1 mg/mL in acetonitrile |
| Formula Weight | 353.76 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation