Learn More
CERILLIANT™ Clomipramine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C1181ML
Description
Clomipramine is a tricyclic antidepressant used to many conditions from major depression and panic disorder to narcolepsy and obsessive compulsion disorder (OCD). This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research.Specifications
| 67-56-1,17321-77-6 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 351.31 | |
| 1 mL |
| C19H24Cl2N2 | |
| 1.0 mg/mL in methanol (as free base) | |
| Ampule |
CERILLIANT is a registered trademark of Cerilliant Corporation