Learn More
CERILLIANT™ Citalopram hydrobromide Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C0951ML
Description
Citalopram is an antidepressant sold under the trade names Celexa™ and Cipramil for the treatment of major depression. Citalopram is a selective serotonin reuptake inhibitor, a class of drugs that also includes fluoxetine, paroxetine, and sertraline. This Certified Snap-N-Spike™ Solution is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research.Specifications
| 67-56-1,59729-32-7 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 405.30 | |
| 1 mL |
| C20H22BrFN2O | |
| 1.0 mg/mL in methanol (as free base) | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation