missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ cis-Tramadol Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing and pharmaceutical research. Tramadol is a synthetic analgesic used to treat moderate to moderately severe pain. The drug has a wide range of applications from treatment of rheumatoid arthritis to fibromyalgia.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Linear Formula | C16H26ClNO2 |
| MDL Number | MFCD03701237 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 1 mL |
| Formula Weight | 299.84 |
CERILLIANT is a registered trademark of Cerilliant Corporation