missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ cis-Tramadol-13C, D3 Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
A stable-labeled internal standard for tramadol testing or isotope dilution methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing, pain prescription monitoring, or forensic analysis. Tramadol is a synthetic analgesic used to treat moderate to moderately severe pain. The drug, sold under trade names Ultram™, Ultracet™, Ralivia, and Tramal, has a wide range of applications from treatment of rheumatoid arthritis to fibromyalgia.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Linear Formula | 13CC15D3H22NO2 · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 1 mL |
| Formula Weight | 303.85 |
CERILLIANT is a registered trademark of Cerilliant Corporation