missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ (+/-)-cis-3-Methylfentanyl Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001668
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001668 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001668 Supplier Merck Emd Millipore Supplier No. M1941EA

Certified reference material

A designer drug and fentanyl analog, (±)-cis-3-Methylfentanyl has an estimated potency 400 to 6000 times greater than morphine. This Certified Spiking Solution™ is suitable for use as starting material in calibrators and controls for (±)-cis-3-Methylfentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications.

Specifications

Packaging Ampule
CAS 78995-18-3
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C23H30N2O · HCl
MDL Number MFCD25974419
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 386.96

CERILLIANT is a registered trademark of Cerilliant Corporation