missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Chlorpromazine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001145
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001145 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001145 Supplier Merck Emd Millipore Supplier No. C9041ML
Only null left

Certified reference material

Chlorpromazine, sold under the trade names Thorazine or Largactil, is a typical antipsychotic used to treat schizophrenia. This Certified Spiking Solution™ is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Specifications

CAS 67-56-1, 69-09-0
Linear Formula C17H19ClN2S · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 355.33
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation