missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Carfentanil-D5 Oxalate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
This stable labeled internal standard is suitable for quantitation of carfentanil in fentanyl GC/MS or LC/MS methods for applications in clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring or isotope dilution. Marketed under the trade name Wildnil; as a general anesthetic for large animals, carfentanil hit the illicit market in 2016 as a new NPS drug or in combination with heroin. Carfentanil, also known as 4-carbomethoxyfentanyl, is a fentanyl analog 10,000 times more potent than morphine, making it one of the most potent synthetic opioids available.This product will be shipped in a can containing one ampoule of C-163-0.5ML due to the compound's acute toxicity.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Concentration | 100 μg/mL in methanol (as free base) |
| Linear Formula | C24H25D5N2O3 · C2H2O4 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 0.5 mL |
| Formula Weight | 489.57 |
CERILLIANT is a registered trademark of Cerilliant Corporation