Learn More
CERILLIANT™ Carbamazepine-10,11-epoxide-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore C1281ML
Catalog No. 01001089
Only null left
Explore available promotions
Description
Description
This stable-labeled internal standard is suitable for use in quantitation of carbamazepine levels or isotope dilution methods by GC/MS or LC/MS for applications in pain prescription monitoring, clinical toxicology, therapeutic drug monitoring or urine drug testing. Carbamazepine-10,11-epoxide is the primary active metabolite of carbamazepine, an anticonvulsant and moodstabilizing drug used primarily in the treatment of epilepsy and neuropathic pain.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol
258.22
1 mL
13C6C9H12N2O2
100 μg/mL in methanol
Ampule
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation