missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Cannabigerolic acid Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001101
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001101 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001101 Supplier Merck Emd Millipore Supplier No. C1421ML

Certified reference material

A non-psychoactive cannabinoid, cannabigerolic acid (CBGA), is the precursor to THCA, CBDA, and CBCA cannabinoids in Cannabis. Cannabigerolic acid reportedly has efficacy in treatment of cancer and schizophrenia. This Certified Spiking Solution™ is suitable for CBGA testing methods by HPLC, LC/MS, or GC/MS for applications in testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK's for Canadian customers.

Specifications

CAS 75-05-8, 25555-57-1
Linear Formula C22H32O4
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Synonym CBGA solution
Formula Weight 360.49
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation