Learn More
CERILLIANT™ Cannabidivarinic Acid (CBDVA) Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C1521ML
Description
Cannabidivarinic acid (CBDVA) is a non-psychoactive cannabinoid found in Cannabis that reportedly has anti-inflammatory properties. This certified Snap-N-Spike™ solution is suitable for CBDVA testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling and pharmaceutical research. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK's for Canadian customers.Specifications
| 75-05-8,31932-13-5 | |
| UN 1648 3 / PGII | |
| 330.42 | |
| 1 mL |
| C20H26O4 | |
| 1.0 mg/mL in acetonitrile | |
| Ampule | |
| 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation