missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Cannabidiol-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore C0841ML
Description
Cannabidiol is a non-psychoactive cannabinoid found in cannabis that has efficacy for a wide range of medical conditions including epilepsy, bipolar disorder, inflammation, and cancer. This stable-labeled internal standard is suitable for quantitation of cannabidiol levels by LC-MS/MS and GC/MS applications for urine drug testing, pharmaceutical research, clinical toxicology or forensic analysis.Specifications
| 67-56-1 | |
| MFCD11044933 | |
| 100 μg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C21H27O2D3 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 317.48 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation