missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ (+/-)-Cannabicyclol (CBL) Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Cannabicyclol (CBL) a non-psychotomimetic cannabinoid found in Cannabis that reportedly exerts therapeutic effects. This Certified Spiking Solution™ is suitable for use in HPLC, GC/MS or LC-MS/MS CBL testing applications in pharmaceutical research, forensic analysis, or Cannabis potency testing & impurity profiling of Cannabis blends or food products.
Specifications
Specifications
| CAS | 75-05-8, 21366-63-2 |
| Linear Formula | C21H30O2 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 314.46 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation