missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Caffeine Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258368
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01258368 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01258368 Supplier Merck Emd Millipore Supplier No. C0511ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Snap-N-Spike™ Reference Solution suitable for use in LC/MS or GC/MS applications from clinical toxicology to dietary supplement testing.

Specifications

Packaging Ampoule
CAS 58-08-2
Product Line CERILLIANT
Physical Form Liquid
Concentration 1 mg/mL in Methanol
MDL Number MFCD00005758
Industry Type Forensics, Toxicology
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation