missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Butyryl fentanyl Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001053
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001053 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001053 Supplier Merck Emd Millipore Supplier No. B0660.5ML
Only null left

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for butyryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Butyryl fentanyl is a designer drug and fentanyl analog that has been associated with numerous overdose deaths around the world. In May 2016, the U.S Drug Enforcement Administration (DEA) classified butyryl fentanyl as a schedule I controlled substance.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C23H30N2O
MDL Number MFCD22054430
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 350.5

CERILLIANT is a registered trademark of Cerilliant Corporation