missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Butabarbital-D5, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore B0651ML
Description
A stable-labeled internal standard suitable for butabarbital testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, forensic analysis or pain prescription monitoring applications. Butabarbital is a barbiturate prescribed as a sleep aid under the trade name Butisol.Specifications
| 67-56-1 | |
| MFCD03428091 | |
| 100 μg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C10H11D5N2O3 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 217.28 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation