Learn More
CERILLIANT™ Buphedrone Ephedrine Metabolite-D3 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore B0511ML
Description
A stable-labeled internal standard suitable for quantitation of Buphedrone Ephedrine Metabolite levels in urine or other matrices by LC/MS or GC/MS for testing applications in clinical toxicology, forensic analysis, or urine drug testing. This bath salt cathinone analog is a primary urinary metabolite of Buphedrone. Buphedrone, also known as MAPB, is a synthetic cathinone and stimulant widely found in numerous bath salt blends across the US, Europe, and Asia.Specifications
| 67-56-1 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 218.74 | |
| 1 mL |
| C11H14D3NO · HCl | |
| 100 μg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) |
CERILLIANT is a registered trademark of Cerilliant Corporation