missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ (+/-)-beta-Hydroxythiofentanyl Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
(±)-β-Hydroxythiofentanyl is an opioid analgesic and fentanyl analog. As of May 2016 (±)-β-Hydroxythiofentanyl is a Schedule I controlled substance in the United States. A Snap-N-Spike™ Reference Solution suitable as starting material for use in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology, urine drug testing or forensic analysis applications.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 100 μg/mL in methanol (as free base) |
| Linear Formula | C20H26N2O2S · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 0.5 mL |
| Formula Weight | 394.96 |
CERILLIANT is a registered trademark of Cerilliant Corporation