missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ beta-Carotene Solution, 100 μg/mL (THF:Ethanol with 0.1% BHT (70:30) (w/v)), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002172
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002172 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002172 Supplier Merck Emd Millipore Supplier No. V0581ML

Certified reference material

β-Carotene, also known as E160a when used as food coloring supplement, is a precursor to vitamin A. Serum levels of β-carotene are an adequate measurement of clinical status and can confirm the diagnosis of carotenoderma and is used to detect fat malabsorption. This Certified Spiking Solution™ is suitable for use as starting material in the preparation of linearity standards, calibrators, or controls in HPLC/UV, LC/MS or GC/MS testing applications such as clinical diagnostic, or food and nutrition testing.

Specifications

CAS 109-99-9, 7235-40-7
Linear Formula C40H56
UN Number UN 1993 - class 3 - PG 2 - RQ - VP ≤ 110kPa - Flammable liquids, n.o.s., HI: all
Concentration 100 μg/mL (THF:ethanol with 0.1% BHT (70:30) (w/v))
Formula Weight 536.87
Packaging Ampule
Quantity 1 mL
Flash Point -21.2°C (-6.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation