missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Baclofen Solution, 500 μg/mL (90:10 MeOH:DMSO), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001057
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001057 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001057 Supplier Merck Emd Millipore Supplier No. B0711ML
Only null left

Certified reference material

A certified solution standard for use in LC/MS or GC/MS applications in clinical toxicology, forensic analysis, pharmaceutical research or diagnostic testing. Baclofen, a derivative of gamma-aminobutyric acid (GABA), is a muscle relaxant used to treat spasticity. Marketed with brand names such as Kemstro, Lioresal or Gablofen, the drug has also shown effectiveness in the treatment of alcoholism. Discontinuation of baclofen can be associated with a withdrawal syndrome resembling benzodiazepine or alcohol withdrawal.

Specifications

CAS 67-56-1
Linear Formula C10H12ClNO2
MDL Number MFCD00055143
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 500 μg/mL (90:10 MeOH:DMSO)
Formula Weight 213.66
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation