missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Aripiprazole Solution, 1.0 mg/mL (50:50 Methanol/Water with 1% 1 N HCl), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001010
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001010 Supplier Merck Emd Millipore Supplier No. A1191ML
Only null left

Certified reference material

A Certified Solution Standard for use in LC or GC applications including forensic drug analysis or calibrator preparation. Aripiprazole is marketed as the atypical antipsychotic Abilify™ for the treatment of schizophrenia, bipolar disorder and clinical depression.

Specifications

CAS 67-56-1
Linear Formula C23H27Cl2N3O2
MDL Number MFCD00892072
UN Number UN 1993C 3 / PGIII
Concentration 1.0 mg/mL (50:50 methanol/water with 1% 1 N HCl)
Formula Weight 448.39
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation