missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Amoxapine, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore A1231ML
Description
Amoxopine is a tetracyclic antidepressant sold under notable trade names including Asendin, Asendis, Defanyl and Demolox for the treatment of major depressive disorder. This Certified Snap-N-Spike™ Solution is suitable for use in LC applications for clinical toxicology, forensic analysis, urine drug testing, pharmaceutical research, or therapeutic drug monitoring.Specifications
| 67-56-1,14028-44-5 | |
| MFCD00069210 | |
| 1.0 mg/mL in methanol | |
| 313.78 | |
| 1 mL |
| C17H16ClN3O | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4]oxazepine | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation