missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ alpha-Pyrrolidinovalerophenone Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001863
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001863 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001863 Supplier Merck Emd Millipore Supplier No. P0901ML
Only null left

Certified reference material

A certified Snap-N-Spike™ solution suitable for use in GC/MS or LC/MS testing methods for bath salt cathinones in clinical toxicology, urine drug testing or forensic analysis applications. Also known as a-PVP or O-2387, a-Pyrrolidinovalerophenone is a derivative of the synthetic cathinones MDPV and pyrovalerone. This illicit drug has stimulant properties and has been found along with MDPV in several bath salt blends across the US, Europe, and Australia.

Specifications

CAS 67-56-1, 5485-65-4
Linear Formula C15H21NO · HCl
MDL Number MFCD28100858
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 267.79
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation