missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ alpha-Pyrrolidinohexanophenone (alpha-PHP HCl) Hydrochloride Solution, 1 mg/mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001901
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001901 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001901 Supplier Merck Emd Millipore Supplier No. P1491ML

Certified reference material

Specifications

CAS 67-56-1, 13415-59-3
Linear Formula C16H23NO · HCl
Concentration 1 mg/mL in methanol (as free base)
Formula Weight 281.82
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation